Learn More
Medchemexpress LLC Guvacoline hydrobrom 1mg | 17210-51-4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1164901MG
Encompass_Preferred
Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid that functions as a weak full agonist at atrial and ileal muscarinic acetylcholine receptors (mAChR). Supplied as a solid research chemical, it is intended for use in receptor pharmacology, in vitro assays, and compound characterization.
  • Hydrobromide salt form suitable for in vitro studies
  • Solid form with specified purity for reproducible results
  • Acts as a muscarinic receptor agonist for pharmacological evaluation
  • Stable under recommended sealed storage conditions to preserve integrity
  • Offered in small quantities tailored to laboratory research needs
Catalog No. 50-002-27261