Learn More
Cayman Chemical GuvacolInhydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 1719010 MG
Encompass_Preferred
A full agonist of atrial and ileal muscarinic receptors with 15-fold reduced potency compared to the related nootropic, arecoline
Catalog No. 50-296-7446