Learn More
Selleck Chemical LLC GV-58-E0137-25MG
SDP
Supplier:  Selleck Chemical LLC E013725MG
Encompass_Preferred
GV-58 is a novel selective N-type and P/Q-type Ca2 channels agonist with IC50s of 7 21 M and 8 81 M respectively
Catalog No. 50-313-8247