Learn More
Trajan Scientific 1/4" GC ferrule (no-hole) 60% Vespel / 40% graphite PK10
SDP
Supplier:  Trajan Scientific 2134XX
Encompass_Preferred
1/4" GC ferrule (no-hole) 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4144