Learn More
Trajan Scientific 1/4" GC ferrule 0.4mm ID 60% Vespel / 40% graphite PK10
SDP
Supplier:  Trajan Scientific 213404
Encompass_Preferred
1/4" GC ferrule 0.4mm ID 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4150