Learn More
Trajan Scientific 1/4" GC ferrule 1/4" ID 60% Vespel / 40% graphite PK500
SDP
Supplier:  Trajan Scientific 213400B
Encompass_Preferred
1/4" GC ferrule 1/4" ID 60% Vespel / 40% graphite PK500
Catalog No. 50-272-4151