Learn More
Trajan Scientific 1/8" GC ferrule 2.4 mm ID 85% Vespel / 15% Graphite PK10
SDP
Supplier:  Trajan Scientific 215224
Encompass_Preferred
1/8" GC ferrule 2.4 mm ID 85% Vespel / 15% Graphite PK10
Catalog No. 50-272-4094