Learn More
Trajan Scientific 1/8" GC ferrule 1/16" ID 60% Vespel / 40% graphite PK10
SDP
Supplier:  Trajan Scientific 213210
Encompass_Preferred
1/8" GC ferrule 1/16" ID 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4156