Learn More
Trajan Scientific 1/16" GC ferrule no-hole (long) 85% Vespel / 15% graphite PK10
SDP
Supplier:  Trajan Scientific 2151XX
Encompass_Preferred
1/16" GC ferrule no-hole (long) 85% Vespel / 15% graphite PK10
Catalog No. 50-272-4100