Learn More
Trajan Scientific 1/16" GC ferrule 0.4 mm ID Agilent style 85% Vespel / 15% graphite PK10
SDP
Supplier:  Trajan Scientific 215164
Encompass_Preferred
1/16" GC ferrule 0.4 mm ID Agilent style 85% Vespel / 15% graphite PK10
Catalog No. 50-272-4105