Learn More
Trajan Scientific 1/16" GC ferrule 0.4 mm ID (long) two-hole 60% Vespel / 40% graphite PK10
SDP
Supplier:  Trajan Scientific 213124
Encompass_Preferred
1/16" GC ferrule 0.4 mm ID (long) two-hole 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4167