Learn More
Trajan Scientific 1/16" GC ferrule 0.3 mm ID (long) three-hole 85% Vespel / 15% Graphite PK10
SDP
Supplier:  Trajan Scientific 215193
Encompass_Preferred
1/16" GC ferrule 0.3 mm ID (long) three-hole 85% Vespel / 15% Graphite PK10
Catalog No. 50-272-4102