Learn More
Trajan Scientific 1/4" GC ferrule (no-hole) 85% Vespel / 15% Graphite PK10
SDP
Supplier:  Trajan Scientific 2154XX
Encompass_Preferred
1/4" GC ferrule (no-hole) 85% Vespel / 15% Graphite PK10
Catalog No. 50-272-4085