Learn More
Medchemexpress LLC HY-119304 1mg Medchemexpress, GW-406381 CAS:221148-46-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1193041MG
Encompass_Preferred
Medchemexpress, HY-119304 1mg GW-406381 CAS:221148-46-5 GW406381, a highly selective cyclooxygenase-2 (COX-2) inhibitor, attenuates spontaneous ectopic discharge in sural nerves of rats following chronic constriction injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9523