Learn More
Eurofins Discoverx GW 4064, 10 mg. (92-1129)
Supplier:  Eurofins Discoverx 921129
Encompass
GW 4064 is a synthetic small molecule which acts as a selective agonist of the orphan nuclear receptor FXR (EC50=15 nM). GW 4064 protects cells and organs against bile acid toxicity, and has been used to elucidate FXR's role in dyslipidemia, diabetes, obesity, and cancer. Has no activity against other nuclear receptors. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9662
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.