Learn More
Abcam GW-441756 hydrochloride, TrKA receptor tyrosine kinase inhibitor, 10MG
Supplier:  Abcam AB14105110MG
Encompass_Preferred
MW 275.3 Da, Purity >99%. Potent, selective TrKA tyrosine kinase inhibitor (IC₅₀ = 2 nM). Displays >100-fold selectivity over a range of other kinases. Antineoplastic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6947