Learn More
Medchemexpress LLC HY-107051 10mg Medchemexpress, GW 766994 CAS:408303-43-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10705110MG
Encompass_Preferred
Medchemexpress, HY-107051 10mg GW 766994 CAS:408303-43-5 GW 766994 is a specific and oral chemokine receptor-3 (CCR3) antagonist, which has entered clinical trial for asthma and eosinophilic bronchitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8363