Learn More
Medchemexpress LLC GW-803430 | 515141-51-2 | 99.1% | 481.99 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY1108350MG
Encompass_Preferred
GW-803430 (GW-3430) is a potent and selective melanin-concentrating hormone receptor 1 (MCH R1) antagonist with a pIC50 of 9.3. It is orally active in an animal model of obesity. For research use only.
  • Potent and selective melanin-concentrating hormone receptor 1 antagonist.
  • Orally active in animal models of obesity.
Catalog No. 50-003-43899