Learn More
TARGETMOL CHEMICALS INC GW 833972A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T643945MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW 833972A is a selective CB2 receptor agonist.GW 833972A has an inhibitory effect on induced neural depolarization and on citric acid-induced cough in an animal model. purity: 98%
Catalog No. 50-257-9361