Learn More
Medchemexpress LLC Gw274150 phosphate | 438542-15-5 | 98.0% | 50 MG

Supplier: Medchemexpress LLC HY12119A50MG
GW274150 phosphate is a selective, orally active inhibitor of inducible nitric oxide synthase (iNOS) supplied as the phosphate salt for research use. It is used in pharmacology studies to probe iNOS-mediated pathways and has reported activity (IC50 ≈ 2.19 μM; Kd ≈ 40 nM).
- Selective inhibitor of inducible nitric oxide synthase (iNOS).
- Reported IC50 2.19 μM and Kd 40 nM.
- High purity (~98.0%) with manufacturer-provided COA.
- Molecular weight 317.30 g·mol⁻¹; formula C8H20N3O6PS.
- Available in small research pack sizes, including 50 MG and solution formats.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.