Learn More
Medchemexpress LLC GW3965 100mg | 405911-09-3 | 100MG
SDP
Supplier:  Medchemexpress LLC HY10627100MG
Encompass_Preferred
GW3965 is a potent selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXR and hLXR respectively[1 [2 [3
Catalog No. 50-002-42892