Learn More
Apexbio Technology LLC GW3965 HCl 405911-17-3 10mg
SDP
Supplier:  Apexbio Technology LLC A844410
Encompass_Preferred
GW3965 HCl is a selective non-steroidal agonist targeting liver X receptors (LXR and LXR ) nuclear transcription factors involved in cholesterol and lipid metabolism Activation of LXRs modulates cholesterol homeostasis by enhancing cholesterol efflux from macrophages stimulating hepatic bile acid synthesis and reducing intestinal cholesterol absorption GW3965 recruits steroid receptor coactivator-1 (SRC-1) to human LXRR in vitro and functions as a full agonist on cellular assays for both LXR and LXR receptors In animal studies oral administration elevated plasma HDL cholesterol levels and increased expression of the cholesterol transporter ABCA1 in macrophages GW3965 is useful in research investigating cholesterol metabolism atherosclerosis and vascular responses mediated by Angiotensin II signaling
Catalog No. 50-000-61889