Learn More
Selleck Chemical LLC GW3965 HCl S2630-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S263010MM/1ML
Encompass_Preferred
GW3965 HCl is a potent selective LXR agonist for hLXR and hLXR with EC50 of 190 and 30 nM in cell-free assays respectively
Catalog No. 50-311-2982