Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GW406108X 100mg | 1644443-92-4 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY115570100MG

Encompass_Preferred

GW406108X is a small-molecule research inhibitor that targets Kif15 (Kinesin-12) and ULK1, reported to inhibit autophagy via ATP-competitive inhibition of ULK1. Supplied as a solid, light yellow to yellow powder for laboratory research use only.

  • Potent autophagy inhibitor: ULK1 pIC50 6.37 (≈427 nM); Kif15 ATPase IC50 0.82 μM.
  • Specified purity: 98.0%.
  • Molecular weight: 400.21 and formula C20H11Cl2NO4.
  • Appearance: light yellow to yellow solid.
  • Recommended storage: powder at -20°C for long term; in solvent, -80°C for up to 6 months.
  • Available in multiple pack sizes including 100 mg.

Catalog No. 50-002-26577


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.