Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GW406108X 10mg | 1644443-92-4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11557010MG

Encompass_Preferred

Small-molecule research inhibitor of Kif15 and ULK1 that blocks autophagic flux and is used in cellular and biochemical studies to probe autophagy and kinesin-related pathways. CAS 1644443-92-4; molecular formula C20H11Cl2NO4; molecular weight 400.21 g/mol.

  • Selective Kif15 ATPase inhibitor (IC50 0.82 μM).
  • ATP-competitive ULK1 inhibitor (pIC50 6.37; ~427 nM).
  • High purity solid for research (98.0% reported by supplier).
  • Molecular formula C20H11Cl2NO4; molecular weight 400.21 g/mol.
  • Recommended storage: powder at -20 °C for long term or 4 °C for short term; in solution store at -80 °C for extended periods.

Catalog No. 50-003-08217


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.