Learn More
Selleck Chemical LLC GW441756 S2891-50mg
SDP
Supplier:  Selleck Chemical LLC S289150MG
Encompass_Preferred
GW441756 is a potent selective inhibitor of TrkA with IC50 of 2 nM with very little activity to c-Raf1 and CDK2 GW441756 produces a relevant increase of caspase-3 that leads to apoptosis
Catalog No. 50-311-1710