Learn More
Medchemexpress LLC GW627368 | 439288-66-1 | 99.9% | 544.62 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1696350MG
Encompass_Preferred
GW627368 is a novel, potent, and selective competitive antagonist of the prostanoid EP4 receptor, demonstrating additional affinity for the human TP receptor. It exhibits pKi values of 7.0 for human prostanoid EP4 and 6.8 for TP receptors. This compound is intended for research use only.
  • Potent and selective competitive antagonist of the prostanoid EP4 receptor.
  • Exhibits additional affinity for the human TP receptor.
  • Inhibits proliferation and invasion of human SUM149 IBC tumor cells.
  • Demonstrates significant tumor regression in vivo.
Catalog No. 50-003-93795