Learn More
Eurofins Discoverx GW7647, 5 mg. (92-1134)
Supplier:  Eurofins Discoverx 921134
Encompass
GW 7647 is a synthetic small molecule agonist of peroxisome proliferator-activated receptor-alpha (PPARα), which is involved in lipid homeostasis regulation. Binds selectively to PPARα over PPARγ and PPARδ, with EC50s of 1-10 nM, ~1 µM, and 1-10 µM respectively. Exhibits anti-inflammatory properties, reduces NO production in macrophages, and has lipid-lowering effects. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9666