Learn More
Medchemexpress LLC HY-13861 5mg , GW7647 CAS:265129-71-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY138615MG
Encompass_Preferred
HY-13861 5mg GW7647 CAS: 265129-71-3 GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9722