Learn More
Medchemexpress LLC GW779439X | 551919-98-3 | 99.9% | 454.45 g/mol | C22H21F3N8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10364550MG
Encompass_Preferred
GW779439X is a pyrazolopyridazine small-molecule research compound reported to inhibit the Stk1 (PASTA) serine/threonine kinase in Staphylococcus aureus and to potentiate β-lactam antibiotics against MRSA. It is supplied as a solid powder for laboratory research use and is characterized by high purity and defined molecular properties.
  • High purity compound suitable for biochemical and microbiological assays.
  • Reported inhibitor of Stk1 kinase that sensitizes MRSA to β-lactam antibiotics.
  • Well-characterized molecular properties: C22H21F3N8, 454.45 g/mol.
  • Stable as a powder when stored under recommended conditions for long-term use.
  • Compatible with common solvents for solution preparation in research workflows.
Catalog No. 50-002-44713