Learn More
Medchemexpress LLC GW806742X hydrochloride | 98.3% | 610.01 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY112292A10MG
Encompass_Preferred
GW806742X hydrochloride is an ATP mimetic and a potent inhibitor of MLKL (Mixed Lineage Kinase Domain-Like protein), binding to its pseudokinase domain with a Kd of 9.3 μM. It also shows activity against VEGFR2 with an IC50 of 2 nM. This compound effectively retards MLKL membrane translocation and inhibits necroptosis.
  • Potent MLKL inhibitor (Kd = 9.3 μM)
  • Activity against VEGFR2 (IC50 = 2 nM)
  • Retards MLKL membrane translocation
  • Inhibits necroptosis
  • Inhibits necroptotic death of wild-type mouse dermal fibroblasts in a dose-dependent manner
  • Inhibits VEGF induced proliferation of HUVECs with an IC50 of 5 nM
Catalog No. 50-004-35918