Learn More
Medchemexpress LLC HY-11003 50mg , GW843682X GW843682 CAS:660868-91-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1100350MG
Encompass_Preferred
Medchemexpress, HY-11003 50mg GW843682X GW843682 CAS:660868-91-7 Formula:C22H18F3N3O4S IC50: 2.2 nM (PLK1), 9.1 nM (PLK3) Purity:98% GW843682X is a selective, ATP-competitive inhibitor of PLK1 and PLK3 , with IC 50 s of 2.2 nM and 9.1 nM, respectively, and is also >100-fold selective against ∼30 other kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3869
 Restricted Item