Learn More
Abcam GYKI 52466, Selective non-competitive AMPA antagonist, 10MG
Supplier:  Abcam AB12033610MG
Encompass_Preferred
MW 329.8 Da, Purity >99%. Selective non-competitive AMPA receptor antagonist (IC₅₀ values are 10-20, approx. 450 and >50 μM for AMPA- , kainate- and NMDA-induced responses, respectively). Skeletal muscle relaxant, orally active anticonvulsant, neuroprotective and anxiolytic in vivo. Also available in simple stock solutions (ab146716) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7666