Learn More
Medchemexpress LLC Gymnemagenin | 22467-07-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN22685MG
Encompass_Preferred
Gymnemagenin is a triterpenoid isolated from *G. sylvestre*. It is an agent for diabetes and obesity and also possesses antiviral properties.
  • Isolated from the plant *Gymnema sylvestre*.
  • Classified as a triterpene.
  • Acts as an agent for diabetes and obesity.
  • Possesses antiviral properties.
  • For research use only.
  • Has poor oral bioavailability in rats.
  • Exhibits short half-life and high clearance in rats.
  • Soluble in DMSO (30 mg/mL) with ultrasonic and warming.
Catalog No. 50-004-21111