Learn More
Medchemexpress LLC Gymnemagenin | 22467-07-8 | MFCD06656306 | 1 MG

Supplier: Medchemexpress LLC HYN22681MG
Gymnemagenin is a triterpenoid isolated from G. sylvestre. It acts as an agent for diabetes and obesity and also exhibits antiviral properties.
- Isolated from G. sylvestre
- Acts as an agent for diabetes and obesity
- Exhibits antiviral properties
- Solid appearance, white to off-white color
- Molecular weight: 506.71
- Formula: C30H50O6
- Structure classification: Terpenoids, Triterpenes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.