Learn More
Selleck Chemical LLC GynostemmapentaphyllumExtract E3343-25MG
SDP
Supplier:  Selleck Chemical LLC E334325MG
Encompass_Preferred
Gynostemma Pentaphyllum Extract isolated from the herb of Gynostemma pentaphyllum enhances exercise performance by promoting myotube differentiation and mitochondrial metabolism through the upregulation of PGC-1 in C2C12 skeletal muscle
Catalog No. 50-000-40550