Learn More
Medchemexpress LLC Gypenoside A | 157752-01-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN24405MG
Encompass_Preferred
Gypenoside A is an orally active triterpenoid compound isolated from Gynostemma pentaphyllum. It possesses anti-inflammatory and antioxidant activities, and offers a protective effect on cardiomyocytes while inhibiting apoptosis. This compound is suitable for research in cardiovascular diseases and inflammation-related conditions.
  • Possesses anti-inflammatory activities
  • Possesses antioxidant activities
  • Offers a protective effect on cardiomyocytes
  • Can inhibit apoptosis
  • Suitable for research in cardiovascular diseases
  • Suitable for research in inflammation-related conditions
  • Reduces secretion of cytokines and chemokines
  • Inhibits ROS production
  • Increases cell viability
  • Activates the AMPK/Foxo1 signaling pathway
Catalog No. 50-004-20544