Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gypenoside A | 157752-01-7 | 99.4% | 899.07 | C46H74O17 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN244010MG

Encompass_Preferred

Gypenoside A is a triterpenoid saponin isolated from Gynostemma pentaphyllum. It exhibits anti-inflammatory and antioxidant activity, shows protective effects on cardiomyocytes, and is used in research on cardiovascular disease and inflammation.

  • High purity: 99.42%.
  • Molecular weight 899.07; formula C46H74O17.
  • CAS number 157752-01-7.
  • White to off-white solid appearance.
  • Storage: store at 4°C and protect from light; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Intended for research and analytical applications in life science studies.

Catalog No. 50-002-89356


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.