Learn More
Medchemexpress LLC Gypenoside XLIX | 94987-08-3 | MFCD22124996 | 99.4% | 1047.23 | C52H86O21 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN199010MG
Encompass_Preferred
Gypenoside XLIX is a dammarane-type glycoside and a selective PPAR-alpha activator isolated from Gynostemma pentaphyllum. Supplied as a research-grade analytical standard, it is intended for laboratory and analytical applications where a high-purity reference compound is required.
  • Dammarane-type glycoside structure found in botanical sources.
  • Selectively activates PPAR-alpha and modulates inflammatory markers in cell assays.
  • High-purity standard suitable for analytical and research use.
  • Solid white to light yellow powder with defined molecular weight and formula.
  • Store under conditions specified on the certificate of analysis to retain stability.
Catalog No. 50-002-96048