Learn More
Gyros Inc GYROLAB AAVX Titer Kit, automated immunoassay, 96 data points, 80 min assay time, 10x less sample vs ELISA, AAV1-8 AAVrh10 compatibility, ready-to-use kit

Supplier: Gyros Inc P0020695
The Gyrolab AAVX Titer Kit from Gyros Protein Technologies offers an automated, nanoliter-scale immunoassay solution for the rapid and precise determination of adeno-associated virus (AAV) vector physical titer. This ready-to-use kit is essential for monitoring critical quality attributes during the process development and manufacturing of AAV viral vectors in cell and gene therapy applications, providing key advantages over traditional ELISA methods.
- Product type: Titer kit
- Application: AAV vector titer determination
- Assay principle: Automated immunoassay
- Serotype compatibility: AAV1-8, AAVrh10
- Serotype incompatibility: AAV9
- Throughput: 96 data points
- Assay time: 80 minutes
- Sample volume: 10x less vs ELISA
- Speed: 4x faster vs ELISA
- Dynamic range: 0.2-1000 ng/mL
- Automated: true
- Format: Ready-to-use kit
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.