Learn More
Medchemexpress LLC Gyrophoric acid | 548-89-0 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN39855MG
Encompass_Preferred
Gyrophoric acid is an ultraviolet filter found in lichen populations and exhibits DPPH radical scavenging activity.
  • Exhibits DPPH radical scavenging activity with an IC50 value of 105.75 μg/ml.
  • Shows antiproliferative activity against human HaCaT cells with an IC50 of 1.7 μM.
  • Inhibits LTB4 biosynthesis in A-23187-stimulated bovine PMNL cells with an IC50 of > 60 μM.
  • Decreases mitochondrial membrane potential in ovarian cancer cells A2780 at 200 μM.
  • Inhibits cell growth and affects expression of Bcl-2, Bax, and Hsp70 proteins in A375 melanoma cancer cells.
  • Induces ROS production, apoptosis, DNA damage, and alters stress/survival protein activity in HeLa cells.
Catalog No. 50-004-18324