Learn More
AdipoGen GYY 4137 (50 mg)
SDP
Supplier:  AdipoGen AGCR13513M050
Encompass_Preferred
Chemical. CAS: 106740-09-4. Formula: C11H16NO2PS2 . C4H9NO. MW: 288.4 . 87.1. Water-soluble, slow-releasing hydrogen sulfide (H2S) donor. Exhibits vasodilator and antihypertensive activity. Anti-inflammatory. Inhibits LPS-induced release of proinflammatory mediators (IL-1beta, IL-6, TNF-alpha, nitric oxide (NO) and PGE2) and increases the synthesis of the anti-inflammatory chemokine IL-10 through NF-kappaB, ATF-2 and HSP27 dependent pathways. Causes stomatal opening and reduces nitric oxide accumulation in plants. Oxidative stress-induced cell death inhibitor. Shows novel anti-cancer effects in vitro and in vivo. Anti-thrombotic via p-selectin dependent mechanism.
Catalog No. 50-205-3892