Learn More
Medchemexpress LLC H-1-Nal-OH | 55516-54-6 | 215.25 | 100 G

Supplier: Medchemexpress LLC HYW008353100G
H-1-Nal-OH is an alanine derivative intended for research use only and is not sold to patients. Amino acids and their derivatives, including H-1-Nal-OH, are utilized as ergogenic supplements known to influence the secretion of anabolic hormones, provide fuel during exercise, improve mental performance under stress, and help prevent muscle damage from exercise.
- Appearance: solid
- Color: white to off-white
- Purity: 99.49%
- Molecular weight: 215.25
- Formula: C13H13NO2
- CAS No.: 55516-54-6
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.