Learn More
Enzo Life Sciences H-1152. dihydrochloride (5mg). CAS: 45146-2-58-1
SDP
Supplier:  Enzo Life Sciences ALX270423M005
Encompass_Preferred
A cell permeable, highly specific, potent and ATP-competitive inhibitor of Rho kinase (ROCK) (Ki=1.6nM). Exhibits a much weaker affinity for other serine/threonine kinases (Ki=630nM for PKA, 9.27mM for PKC and 10.1mM for MLCK). More potent and selective than Y-27632 (ALX-270-333). Inhibits the phosphorylation of MARCKS in cells stimulated by lysophosphatidic acid. Inhibits EP3-stimulated NO formation. Prevents fragmentation of apoptotic cells. Relieves neuropathic pain. Alternative name: H-1152P . 2HCl, (S)-(+)-2-Methyl-1-[(4-methyl-5-isoquinolinyl)sulfonyl]homopiperazine . 2HCl. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8546