Learn More
Cayman Chemical H-151, 941987-60-6, MFCD08026581, 10mg
SDP
Supplier:  Cayman Chemical 2585710
Encompass_Preferred
Molecular Formula: C17H17N3O, Purity: ≥98%, Formula Weight: 279.3, Formal Name: N-(4-ethylphenyl)-N'-1H-indol-3-yl-urea.
Catalog No. NC1622480