Learn More
Selleck Chemical LLC H-151
SDP
Supplier:  Selleck Chemical LLC S66521G
Encompass_Preferred
H-151 is a highly potent and covalent inhibitor of STING that has noteworthy inhibitory activity both in human cells and in vivo
Catalog No. 50-000-02694