Learn More
Medchemexpress LLC H-151 | 941987-60-6 | 99.86% | 279.34 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11269310MM/1ML
Encompass_Preferred
H-151 is a potent, selective and covalent antagonist of STING that has noteworthy inhibitory activity both in cells and in vivo. H-151 reduces TBK1 phosphorylation and suppresses STING palmitoylation. It can be used for the research of autoinflammatory disease.
  • Potent, selective, and covalent antagonist of STING.
  • Exhibits noteworthy inhibitory activity in cells and in vivo.
  • Reduces TBK1 phosphorylation.
  • Suppresses STING palmitoylation.
  • Useful for research of autoinflammatory disease.
  • Reduces IFNβ luciferase reporter measurements.
  • Inhibits phosphorylation of TBK1.
  • Suppresses palmitoylation.
  • Markedly reduces systemic cytokine responses.
  • Exhibits notable efficacy in certain mouse models.
  • Reaches effective systemic levels.
  • Displays a short half-life in serum.
  • Forms an adduct to mmSTING.
Catalog No. 50-004-37873