Learn More
Medchemexpress LLC H-151 Alkyne | 2244892-03-1 | 95.1% | 275.30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY176566
Encompass_Preferred
H-151 Alkyne (Compound H-151-AL) is a selective STING (Stimulator of interferon genes) inhibitor. It is promising for research of autoimmune diseases such as systemic lupus erythematosus, scleroderma, and autoinflammatory diseases.
  • Selective STING (Stimulator of interferon genes) inhibitor.
  • Promising for research of autoimmune diseases such as systemic lupus erythematosus, scleroderma, and autoinflammatory diseases.
Catalog No. 50-003-84101