Learn More
Medchemexpress LLC H-2-Nal-OH | 58438-03-2 | 215.25 | 5 G

Supplier: Medchemexpress LLC HYW0083795G
H-2-Nal-OH is an alanine derivative intended for research use only. Amino acids and their derivatives, like H-2-Nal-OH, are recognized for their beneficial properties as ergogenic dietary substances. They can influence the secretion of anabolic hormones, provide fuel during exercise, improve mental performance during stress-related tasks, and help prevent exercise-induced muscle damage.
- Appearance: Solid
- Color: White to off-white
- Solubility: 3.33 mg/mL in H2O (15.47 mM; with ultrasonic and adjustment of pH to 12 with NaOH)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.