Learn More
Selleck Chemical LLC H 33258 trihydrochloride E2910-5MG
SDP
Supplier:  Selleck Chemical LLC E29105MG
Encompass_Preferred
bisBenzimide H 33258 Trihydrochloride(Hoechst 33258 trihydrochloride H 33258 trihydrochloride) is a marker dye in Hoechst series Hoechst is A live nuclear marker dye It binds to the grooves in the DNA double strand which tends to be A/ T-rich DNA strand Although it binds to all nucleic acids the A/ T-rich double strand DNA significantly enhances fluorescence intensity Therefore Hoechst dye can be used for living cell labeling The fluorescence intensity of Hoechst dye increases with the increase of pH of solution It exhibits anti-tumor activity IC50 of 51 31 4 56 M in HeLa cells
Catalog No. 50-000-39682